Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-4-methylthiophenylboronic acid pinacol ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

31014-1G 31014-25G 31014-500MG 31014-5G
76901-834EA 409.51 USD InStock
76901-834 76901-836 76901-838 76901-840
2-Methyl-4-methylthiophenylboronic acid pinacol ester ≥97% research grade
2-Methyl-4-methylthiophenylboronic acid pinacol ester


Formula: C₁₄H₂₁BO₂S
Storage Temperature: Ambient
MDL Number: MFCD29088246
CAS Number: 1899905-99-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty