Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-(tert-butyl)-1-iodobenzene ≥97% research grade

Supplier: AOB CHEM USA
Warning

31309-1G 31309-25G 31309-5G
76901-948EA 102.91 USD InStock
76901-948 76901-950 76901-952
2-Bromo-4-(tert-butyl)-1-iodobenzene ≥97% research grade
2-Bromo-4-(tert-butyl)-1-iodobenzene


Formula: C₁₀H₁₂BrI
MW: 339.01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09878146
CAS Number: 860435-39-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty