Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Difluoro-5-methoxybenzaldehyde ≥97% research grade

Supplier: AOB CHEM USA
Warning

31727-1G 31727-25G 31727-500MG 31727-5G
76902-060EA 124.13 USD InStock
76902-060 76902-062 76902-064 76902-066
2,4-Difluoro-5-methoxybenzaldehyde ≥97% research grade
2,4-Difluoro-5-methoxybenzaldehyde


Formula: C₈H₆F₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD24539280
CAS Number: 177034-25-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty