Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethoxy-4-(trifluoromethoxy)phenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

31737-1G 31737-500MG 31737-5G
76902-076EA 530.45 USD InStock
76902-076 76902-078 76902-080
2-Ethoxy-4-(trifluoromethoxy)phenylboronic acid ≥97% research grade
2-Ethoxy-4-(trifluoromethoxy)phenylboronic acid


Formula: C₉H₁₀BF₃O₄
Storage Temperature: Refrigerator
MDL Number: MFCD28384206
CAS Number: 1704064-18-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty