Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-3-(N,N-dimethylaminocarbonyl)phenylboronic acid pinacol ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

32530-1G 32530-25G 32530-500MG 32530-5G
76902-358EA 350.1 USD InStock
76902-358 76902-360 76902-362 76902-364
2-Fluoro-3-(N,N-dimethylaminocarbonyl)phenylboronic acid pinacol ester ≥97% research grade
2-Fluoro-3-(N,N-dimethylaminocarbonyl)phenylboronic acid pinacol ester


Formula: C₁₅H₂₁BFNO₃
MW: 293.142 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18730342
CAS Number: 2121512-58-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty