Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dichloro-3-methoxybenzaldehyde ≥95% research grade

Supplier: AOB CHEM USA
Warning

33590-1G 33590-25G 33590-500MG 33590-5G
76902-620EA 350.1 USD InStock
76902-620 76902-622 76902-624 76902-626
2,5-Dichloro-3-methoxybenzaldehyde ≥95% research grade
2,5-Dichloro-3-methoxybenzaldehyde


Formula: C₈H₆Cl₂O₂
MW: 205.038 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31382556
CAS Number: 1629253-43-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty