Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-3-isopropylbenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

34271-25G 34271-5G
76902-834EA 584.56 USD InStock
76902-834 76902-836
2-Hydroxy-3-isopropylbenzoic acid ≥97% research grade
2-Hydroxy-3-isopropylbenzoic acid


Formula: C₁₀H₁₂O₃
MW: 180.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00075272
CAS Number: 7053-88-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty