Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-3-bromo-5-carboxyphenylboronic acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

34294-1G 34294-25G 34294-500MG 34294-5G
76902-846EA 138.98 USD InStock
76902-846 76902-848 76902-850 76902-852
2-Fluoro-3-bromo-5-carboxyphenylboronic acid ≥95% research grade
2-Fluoro-3-bromo-5-carboxyphenylboronic acid


Formula: C₇H₅BBrFO₄
Storage Temperature: Refrigerator
MDL Number: MFCD12026723
CAS Number: 1452575-84-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty