Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Phenylpyridin-3-yl)boronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

34506-1G 34506-25G 34506-500MG 34506-5G
76902-898EA 381.93 USD InStock
76902-898 76902-900 76902-902 76902-904
(2-Phenylpyridin-3-yl)boronic acid ≥97% research grade
(2-Phenylpyridin-3-yl)boronic acid


Formula: C₁₁H₁₀BNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD07438014
CAS Number: 1029654-14-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty