Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4,5-dichlorobenzaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

35894-1G 35894-500MG 35894-5G 35894-25G
76903-222EA 553.79 USD InStock
76903-222 76903-224 76903-226 77655-380
2-Bromo-4,5-dichlorobenzaldehyde ≥97%
2-Bromo-4,5-dichlorobenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₃BrCl₂O
Storage Temperature: Refrigerator
MDL Number: MFCD32203989
CAS Number: 1259188-29-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty