Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-(1,3-oxazol-5-yl)pyridine ≥97%

Supplier: AOB CHEM USA
Warning

36095-1G 36095-25G 36095-500MG 36095-5G
76903-236EA 350.1 USD InStock
76903-236 76903-238 76903-240 76903-242
2-Chloro-5-(1,3-oxazol-5-yl)pyridine ≥97%
2-Chloro-5-(1,3-oxazol-5-yl)pyridine
Precaución: for Research Use Only


Formula: C₈H₅ClN₂O
MW: 180.591 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05974999
CAS Number: 860648-90-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty