Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-3-methyl-5-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOB CHEM USA
Warning

36362-1G 36362-25G 36362-500MG 36362-5G
76903-262EA 254.62 USD InStock
76903-262 76903-264 76903-266 76903-268
2-Fluoro-3-methyl-5-(trifluoromethyl)benzaldehyde ≥97%
2-Fluoro-3-methyl-5-(trifluoromethyl)benzaldehyde
Precaución: for Research Use Only


Formula: C₉H₆F₄O
Storage Temperature: Refrigerator
MDL Number: MFCD31645674
CAS Number: 2055275-09-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty