Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-6-(difluoromethoxy)naphthalene ≥97%

Supplier: AOB CHEM USA
Warning

36689-1G 36689-25G 36689-500MG 36689-5G
76903-330EA 315.09 USD InStock
76903-330 76903-332 76903-334 76903-336
2-Bromo-6-(difluoromethoxy)naphthalene ≥97%
2-Bromo-6-(difluoromethoxy)naphthalene
Precaución: for Research Use Only


Formula: C₁₁H₇BrF₂O
MW: 273.073 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16040753
CAS Number: 244103-47-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty