Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dichloro-4-hydroxybenzyl alcohol ≥95% research grade

Supplier: AOB CHEM USA
Warning

38345-1G 38345-25G 38345-500MG 38345-5G
76903-634EA 257.8 USD InStock
76903-634 76903-636 76903-638 76903-640
3,5-Dichloro-4-hydroxybenzyl alcohol ≥95% research grade
3,5-Dichloro-4-hydroxybenzyl alcohol


Formula: C₇H₆Cl₂O₂
MW: 193.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06202847
CAS Number: 22002-17-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty