Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Benzyloxy-5-pyrimidineboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

39453-1G 39453-25G 39453-500MG 39453-5G
76903-770EA 236.59 USD InStock
76903-770 76903-772 76903-774 76903-776
2-Benzyloxy-5-pyrimidineboronic acid ≥97% research grade
2-Benzyloxy-5-pyrimidineboronic acid


Formula: C₁₁H₁₁BN₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD07375141
CAS Number: 1217500-86-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty