Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Ethoxy-4-(trifluoromethyl)phenyl)boronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

39775-1G 39775-25G 39775-500MG 39775-5G
76903-786EA 215.37 USD InStock
76903-786 76903-788 76903-790 76903-792
(2-Ethoxy-4-(trifluoromethyl)phenyl)boronic acid ≥97% research grade
(2-Ethoxy-4-(trifluoromethyl)phenyl)boronic acid


Formula: C₉H₁₀BF₃O₃
MW: 233.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18087684
CAS Number: 1260518-74-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty