Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-methylpyridine-4-boronic acid ≥97%

Supplier: AOB CHEM USA
Warning

44128-1G 44128-25G 44128-500MG 44128-5G
76903-938EA 381.93 USD InStock
76903-938 76903-940 76903-942 76903-944
2-Chloro-5-methylpyridine-4-boronic acid ≥97%
2-Chloro-5-methylpyridine-4-boronic acid
Precaución: for Research Use Only


Formula: C₆H₇BClNO₂
Storage Temperature: Ambient
MDL Number: MFCD16610704
CAS Number: 2377605-91-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty