Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-methyl-6-nitroaniline ≥97%

Supplier: AOB CHEM USA
Warning

45564-1G 45564-25G 45564-500MG 45564-5G
76903-982EA 124.13 USD InStock
76903-982 76903-984 76903-986 76903-988
2-Chloro-4-methyl-6-nitroaniline ≥97%
2-Chloro-4-methyl-6-nitroaniline


Formula: C₇H₇ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD08059084
CAS Number: 5465-33-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty