Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methoxy-4-phenylbenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

46307-1G 46307-25G 46307-500MG 46307-5G
76904-062EA 363.89 USD InStock
76904-062 76904-064 76904-066 76904-068
3-Methoxy-4-phenylbenzoic acid ≥97%
3-Methoxy-4-phenylbenzoic acid
Precaución: for Research Use Only


Formula: C₁₄H₁₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD06203597
CAS Number: 175153-20-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty