Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-5-methylnicotinic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

48213-1G 48213-25G 48213-500MG 48213-5G
76904-228EA 409.51 USD InStock
76904-228 76904-230 76904-232 76904-234
2-Methoxy-5-methylnicotinic acid ≥97% research grade
2-Methoxy-5-methylnicotinic acid


Formula: C₈H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD15530274
CAS Number: 1227594-72-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty