Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-6-(tert-butyl)phenol ≥97%

Supplier: AOB CHEM USA
Warning

48444-1G 48444-25G 48444-500MG 48444-5G
76904-322EA 363.89 USD InStock
76904-322 76904-324 76904-326 76904-328
2-Bromo-6-(tert-butyl)phenol ≥97%
2-Bromo-6-(tert-butyl)phenol
Precaución: for Research Use Only


Formula: C₁₀H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD16996888
CAS Number: 23159-87-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty