Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-chloro-6-fluorobenzoic acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

49566-1G 49566-500MG 49566-5G 49566-25G
76904-492EA 468.92 USD InStock
76904-492 76904-494 76904-496 77659-824
2-Bromo-4-chloro-6-fluorobenzoic acid ≥95% research grade
2-Bromo-4-chloro-6-fluorobenzoic acid


Formula: C₇H₃BrClFO₂
MW: 253.453 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11845467
CAS Number: 1082040-61-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty