Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-1,3-difluoro-5-iodobenzene ≥95%

Supplier: AOB CHEM USA
Warning

60434-1G 60434-25G 60434-500MG 60434-5G
76905-406EA 132.62 USD InStock
76905-406 76905-408 76905-410 76905-412
2-Chloro-1,3-difluoro-5-iodobenzene ≥95%
2-Chloro-1,3-difluoro-5-iodobenzene
Precaución: for Research Use Only


Formula: C₆H₂ClF₂I
Storage Temperature: Ambient
MDL Number: MFCD18415606
CAS Number: 1261685-21-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty