Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dichloro-3-methylbenzoic acid ≥95%

Supplier: AOB CHEM USA
Warning

69236-1G 69236-25G 69236-500MG 69236-5G
76905-758EA 233.4 USD InStock
76905-758 76905-760 76905-762 76905-764
2,5-Dichloro-3-methylbenzoic acid ≥95%
2,5-Dichloro-3-methylbenzoic acid
Precaución: for Research Use Only


Formula: C₈H₆Cl₂O₂
MW: 205.038 g/mol
Storage Temperature: Ambient
CAS Number: 1160573-37-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty