Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-4-phenoxybenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

83416-1G 83416-25G 83416-500MG 83416-5G
76905-990EA 381.93 USD InStock
76905-990 76905-992 76905-994 76905-996
2-Methoxy-4-phenoxybenzoic acid ≥97%
2-Methoxy-4-phenoxybenzoic acid
Precaución: for Research Use Only


Formula: C₁₄H₁₂O₄
Storage Temperature: Ambient
CAS Number: 95420-77-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty