Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluoro-5-formylnicotinic acid ≥97%

Supplier: AOB CHEM USA
Warning

91877-1G 91877-25G 91877-500MG 91877-5G
76906-080EA 131.56 USD InStock
76906-080 76906-082 76906-084 76906-086
2,6-Difluoro-5-formylnicotinic acid ≥97%
2,6-Difluoro-5-formylnicotinic acid
Precaución: for Research Use Only


Formula: C₇H₃F₂NO₃
MW: 187.1 g/mol
Storage Temperature: Ambient
CAS Number: 2921838-91-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty