Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2,6-difluorophenylboronic acid ≥95%

Supplier: AOB CHEM USA
Warning

11260-1G 11260-25G 11260-5G
76924-516EA 117.76 USD InStock
76924-516 76924-518 76924-520
4-Chloro-2,6-difluorophenylboronic acid ≥95%
4-Chloro-2,6-difluorophenylboronic acid
Precaución: for Research Use Only


Formula: C₆H₄BClF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03095138
CAS Number: 925911-61-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty