Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methyl-2-nitrophenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

12166-1G 12166-25G 12166-500MG 12166-5G
76924-712EA 260.99 USD InStock
76924-712 76924-714 76924-716 76924-718
4-Methyl-2-nitrophenylboronic acid ≥97%
4-Methyl-2-nitrophenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₈BNO₄
Storage Temperature: Ambient
MDL Number: MFCD06659821
CAS Number: 143697-03-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty