Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Methoxycarbonyl)pyridine-3-boronic acid, pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

13460-1G 13460-25G 13460-5G
76924-870EA 70.02 USD InStock
76924-870 76924-872 76924-874
5-(Methoxycarbonyl)pyridine-3-boronic acid, pinacol ester ≥97%
5-(Methoxycarbonyl)pyridine-3-boronic acid, pinacol ester
Precaución: for Research Use Only


Formula: C₁₃H₁₈BNO₄
Storage Temperature: Refrigerator
MDL Number: MFCD09027079
CAS Number: 1025718-91-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty