Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxy-2-nitrophenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

13833-1G 13833-25G 13833-500MG 13833-5G
76924-908EA 439.22 USD InStock
76924-908 76924-910 76924-912 76924-914
4-Methoxy-2-nitrophenylboronic acid ≥97%
4-Methoxy-2-nitrophenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₈BNO₅
Storage Temperature: Refrigerator
MDL Number: MFCD09878340
CAS Number: 860034-09-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty