Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-hydroxybenzeneboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

14005-1G 14005-25G 14005-5G
76924-924EA 66.84 USD InStock
76924-924 76924-926 76924-928
5-Bromo-2-hydroxybenzeneboronic acid ≥97% research grade
5-Bromo-2-hydroxybenzeneboronic acid


Formula: C₆H₆BBrO₃
Storage Temperature: Refrigerator
MDL Number: MFCD00093408
CAS Number: 89598-97-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty