Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Formylpyridine-3-boronic acid ≥97%

Supplier: AOB CHEM USA
Warning

14139-1G 14139-25G 14139-500MG 14139-5G
76925-100EA 260.99 USD InStock
76925-100 76925-102 76925-104 76925-106
5-Formylpyridine-3-boronic acid ≥97%
5-Formylpyridine-3-boronic acid
Precaución: for Research Use Only


Formula: C₆H₆BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD07374886
CAS Number: 919347-69-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty