Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(5-Fluoro-2-(methoxymethoxy)phenyl)boronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

14178-25G 14178-5G
76925-118EA 332.07 USD InStock
76925-118 76925-120
(5-Fluoro-2-(methoxymethoxy)phenyl)boronic acid ≥97% research grade
(5-Fluoro-2-(methoxymethoxy)phenyl)boronic acid


Formula: C₈H₁₀BFO₄
Storage Temperature: Refrigerator
MDL Number: MFCD11044924
CAS Number: 488713-34-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty