Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-cyanobenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

16875-1G 16875-25G 16875-500MG 16875-5G
76925-940EA 159.14 USD InStock
76925-940 76925-942 76925-944 76925-946
5-Bromo-2-cyanobenzoic acid ≥97%
5-Bromo-2-cyanobenzoic acid
Precaución: for Research Use Only


Formula: C₈H₄BrNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD15523585
CAS Number: 1032231-28-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty