Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methylthiophene-3-boronic acid pinacol ester ≥95% research grade

Supplier: AOB CHEM USA
Warning

19139-1G 19139-25G 19139-5G
76926-680EA 62.6 USD InStock
76926-680 76926-682 76926-684
4-Methylthiophene-3-boronic acid pinacol ester ≥95% research grade
4-Methylthiophene-3-boronic acid pinacol ester


Formula: C₁₁H₁₇BO₂S
Storage Temperature: Ambient
MDL Number: MFCD09265133
CAS Number: 476620-20-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty