Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Carboxy-3-isopropylphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

19500-1G 19500-500MG 19500-5G
76926-772EA 831.75 USD InStock
76926-772 76926-774 76926-776
4-Carboxy-3-isopropylphenylboronic acid ≥97%
4-Carboxy-3-isopropylphenylboronic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₃BO₄
Storage Temperature: Refrigerator
MDL Number: MFCD23379523
CAS Number: 851335-14-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty