Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Benzyloxy)-2-methylphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

20008-1G 20008-25G 20008-500MG 20008-5G
76926-956EA 135.8 USD InStock
76926-956 76926-958 76926-960 76926-962
5-(Benzyloxy)-2-methylphenylboronic acid ≥97%
5-(Benzyloxy)-2-methylphenylboronic acid
Precaución: for Research Use Only


Formula: C₁₄H₁₅BO₃
MW: 242.078 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17011987
CAS Number: 1451391-56-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty