Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-2-(methylthio)benzoic acid ≥95%

Supplier: AOB CHEM USA
Warning

20136-1G 20136-25G 20136-500MG 20136-5G
76927-012EA 185.66 USD InStock
76927-012 76927-014 76927-016 76927-018
5-Fluoro-2-(methylthio)benzoic acid ≥95%
5-Fluoro-2-(methylthio)benzoic acid
Precaución: for Research Use Only


Formula: C₈H₇FO₂S
MW: 186.203 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18860450
CAS Number: 1256727-01-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty