Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-hydroxyphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

20606-1G 20606-25G 20606-500MG 20606-5G
76927-186EA 201.58 USD InStock
76927-186 76927-188 76927-190 76927-192
4-Bromo-2-hydroxyphenylboronic acid ≥97%
4-Bromo-2-hydroxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₆H₆BBrO₃
Storage Temperature: Refrigerator
MDL Number: MFCD18447611
CAS Number: 1451393-43-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty