Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 5-chloro-2-fluoronicotinate ≥97% research grade

Supplier: AOB CHEM USA
Warning

21052-1G 21052-25G 21052-500MG 21052-5G
76927-322EA 245.07 USD InStock
76927-322 76927-324 76927-326 76927-328
Ethyl 5-chloro-2-fluoronicotinate ≥97% research grade
Ethyl 5-chloro-2-fluoronicotinate


Formula: C₈H₇ClFNO₂
MW: 203.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698189
CAS Number: 1214345-81-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty