Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-fluoro-N,N-dimethylnicotinamide ≥97%

Supplier: AOB CHEM USA
Warning

22064-1G 22064-25G 22064-500MG 22064-5G
76927-580EA 260.99 USD InStock
76927-580 76927-582 76927-584 76927-586
5-Bromo-2-fluoro-N,N-dimethylnicotinamide ≥97%
5-Bromo-2-fluoro-N,N-dimethylnicotinamide
Precaución: for Research Use Only


Formula: C₈H₈BrFN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD19105403
CAS Number: 1310416-60-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty