Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-7-methoxy-1H-indazole ≥97%

Supplier: AOB CHEM USA
Warning

22169-1G 22169-25G 22169-500MG 22169-5G
76927-620EA 180.36 USD InStock
76927-620 76927-622 76927-624 76927-626
4-Bromo-7-methoxy-1H-indazole ≥97%
4-Bromo-7-methoxy-1H-indazole


Formula: C₈H₇BrN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD17016112
CAS Number: 938062-01-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty