Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-3,5-dimethylbenzaldehyde ≥97%

Supplier: AOB CHEM USA

22813-25G 22813-5G
76927-888EA 310.85 USD InStock
76927-888 76927-890
4-Fluoro-3,5-dimethylbenzaldehyde ≥97%
4-Fluoro-3,5-dimethylbenzaldehyde


Formula: C₉H₉FO
MDL Number: MFCD06660264
CAS Number: 363134-35-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty