Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Cyano-2,6-dimethylphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

23065-1G 23065-25G 23065-500MG 23065-5G
76927-924EA 260.99 USD InStock
76927-924 76927-926 76927-928 76927-930
4-Cyano-2,6-dimethylphenylboronic acid ≥97%
4-Cyano-2,6-dimethylphenylboronic acid
Precaución: for Research Use Only


Formula: C₉H₁₀BNO₂
Storage Temperature: Ambient
MDL Number: MFCD22056751
CAS Number: 1451391-43-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty