Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Benzyloxy)-2-fluoropyridine ≥97% research grade

Supplier: AOB CHEM USA
Warning

23231-1G 23231-25G 23231-500MG 23231-5G
76927-964EA 155.96 USD InStock
76927-964 76927-966 76927-968 76927-970
5-(Benzyloxy)-2-fluoropyridine ≥97% research grade
5-(Benzyloxy)-2-fluoropyridine


Formula: C₁₂H₁₀FNO
Storage Temperature: Ambient
MDL Number: MFCD24171051
CAS Number: 1204483-95-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty