Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-(methylsulfanyl)phenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

24620-1G 24620-25G 24620-500MG 24620-5G
76928-462EA 185.66 USD InStock
76928-462 76928-464 76928-466 76928-468
5-Chloro-2-(methylsulfanyl)phenylboronic acid ≥97% research grade
5-Chloro-2-(methylsulfanyl)phenylboronic acid


Formula: C₇H₈BClO₂S
MW: 202.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22543736
CAS Number: 1451392-55-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty