Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3,5-dimethylbenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

25510-1G 25510-25G 25510-5G
76928-792EA 73.21 USD InStock
76928-792 76928-794 76928-796
4-Bromo-3,5-dimethylbenzoic acid ≥97%
4-Bromo-3,5-dimethylbenzoic acid


Formula: C₉H₉BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD16250111
CAS Number: 7697-32-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty