Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-methoxybenzyl alcohol ≥97%

Supplier: AOB CHEM USA
Warning

26081-1G 26081-25G 26081-500MG 26081-5G
76929-038EA 135.8 USD InStock
76929-038 76929-040 76929-042 76929-044
5-Chloro-2-methoxybenzyl alcohol ≥97%
5-Chloro-2-methoxybenzyl alcohol
Precaución: for Research Use Only


Formula: C₈H₉ClO₂
MW: 172.609 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00086350
CAS Number: 7035-10-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty