Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-3-hydroxy-5-methylphenylboronic acid, pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

26225-1G 26225-25G 26225-500MG 26225-5G
76929-112EA 350.1 USD InStock
76929-112 76929-114 76929-116 76929-118
4-Fluoro-3-hydroxy-5-methylphenylboronic acid, pinacol ester ≥97%
4-Fluoro-3-hydroxy-5-methylphenylboronic acid, pinacol ester
Precaución: for Research Use Only


Formula: C₁₃H₁₈BFO₃
MW: 252.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045267
CAS Number: 2121512-07-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty