Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chlorobiphenyl-3-ylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

27447-1G 27447-25G 27447-500MG 27447-5G
76929-494EA 381.93 USD InStock
76929-494 76929-496 76929-498 76929-500
5-Chlorobiphenyl-3-ylboronic acid ≥97%
5-Chlorobiphenyl-3-ylboronic acid
Precaución: for Research Use Only


Formula: C₁₂H₁₀BClO₂
Storage Temperature: Ambient
MDL Number: MFCD18384165
CAS Number: 1186403-21-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty